Learn More
Brand™ HDPE Scoops
Product Code. 10049890 Shop All Brand Products
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Length (Metric):
350 mm
400 mm
Unit Size:
Pack of 6
Product Code. 10049890
Brand: Brand™ 139040
Scoop powdered and granular material with ease. BRAND™ HDPE Scoops provide sturdy handling with durable polyethylene.
  • Manufactured from HDPE
  • Natural colored
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 1000 mL
Color Natural
Product Type Scoop
Material HDPE
Length (Metric) 400 mm