Learn More
VITLAB™ HDPE Measuring Scoop
Product Code. 10106550
Click to view available options
Capacity (Metric):
15 mL
65 mL
110 mL
350 mL
Unit Size:
Each
Pack of 12
Pack of 6
Product Code. 10106550
Brand: VITLAB™ 40093
Only null left
Add to basket
Add to basket
Use these corrosion resistant scoops for applications in the food industry. VITLAB™ HDPE Measuring Scoops are lighter than metal, don’t dent and feature good chemical resistance.
  • Conical in shape with tapered filling edge
Specifications
Capacity (Metric) 15 mL
Color Natural
Product Type Measuring Scoop
For Use With (Application) Suitable as weighing
Material HDPE