Learn More
Kartell™ Tank with Spigot
Product Code. p-8007239
Change view
Click to view available options
Capacity (Metric):
10 L
20 L
Unit Size:
Pack of 4
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10079922 10 L Pack of 5
10640204 20 L Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10079922 Supplier Kartell™ Supplier No. 0177000
Only null left
Add to basket
Add to basket
Designed to rationalize laboratory and small production line organization. Kartell™ Tank with Spigot gives the opportunity to optimize water, aqueous solutions and solvents availability in a reduced space.
  • Made of food grade High Density PE
  • Includes comfortable handle for displacement
  • The spigot is placed in a position allowing the tank to stand, as to facilitate the use of these tanks in packaging and transporting liquids
  • Suitable for foodstuff
Specifications
Material HDPE
Capacity (Metric) 10 L
Length (Metric) 380 mm
Width (Metric) 300 mm
Height (Metric) 160 mm
No. per Pack 5