Learn More
SymLine Safety Waste Caps
GREENER_CHOICE
Product Code. 17172566
Click to view available options
Size:
GL 45, GL 25
GL45
S60/61
Includes:
2 x PFA-fittings
4 x PFA-fittings
4 x PFA-fittings, 1 x tube connector
7 x PFA-fittings
Electronic fill level control
Filling Lance, Connector for exhaust filter
Unit Size:
Each
Product Code. 17172566
Brand: SymLine 306489
Safety Waste Caps safely connect the pipework to the waste container. The integrated exhaust filter blocks solvent vapours – for the protection of health and the environment.
  • Safety Waste Caps are available with different thread sizes for attachment to the pipe system and with different numbers of hose connections for disposal of the liquid waste.
  • Optionally, Safety Waste Caps are available with electronic level sensors that indicate the degree of filling of the disposal container.
  • By using a 4-in-1 collector (306509), up to 4 Safety Waste Caps can be connected simultaneously to only one connection point on the pipe system
  • Specifications
    For Use With (Equipment) SymLine System for capillaries with (OD) 1.6/2.3/3.2 mm
    Includes 2 x PFA-fittings
    Material PTFE
    Size GL45
    For Use With (Application) Waste Disposal System
    Type Safety Waste Cap