Learn More
Verenigde Kunststof Bedrijven™ HDPE Barrel
Product Code. 10257354
Change view
Click to view available options
Capacity (Metric):
30 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10257354 30 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10257354 Supplier Verenigde Kunststof Bedrijven™ Supplier No. 602900
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Verenigde Kunststof Bedrijven™ HDPE Barrel is designed for storage, handling and transportation of products manufactured by the chemical, petrochemical and food industries.
  • HDPE UN approved Lid barrels
  • Removable black Lid
  • Metal fastener
  • For transport and storage Chemical, Petrochemical, Food Industries
  • Complete Draining
  • enabling re-use or recycling
  • Does not age or corrode
  • Easy to Stack and pallatise
Specifications
Certifications/Compliance UN Approved
Product Type Barrel
Closure Type Removable Lid
Capacity (Metric) 30 L
Color Blue
Material HDPE
Includes Deckel
Quantity 6
Shape Pallet
Length (Metric) 350 mm
Height (Metric) 461 mm
Diameter (Metric) Opening 250 mm