Learn More
R&D Systems™ HCoV-229E Nucleocapsid (NP_073556.1) VersaClone cDNA
Specifications
Specifications
| Gene | Nucleocapsid |
| Accession Number | NP_073556.1 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1183 bp |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | HCoV-229 E |
| Quantity | 10 μg |