Learn More
Tocris Bioscience™ HC 067047
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
HC 067047 is a potent and selective TRPV4 antagonist. Reversibly inhibits currents through mouse, human and rat TRPV4 orthologs (IC50 values are 17, 48 and 133 nM respectively). Also inhibits the endogenous TRPV4-mediated response to 4α-PDH (IC50 = 22 nM). Selective for TRPV4 over TRPV1, TRPV2, TRPV3 and TRPM8 channels.
Specifications
Specifications
| Chemical Name or Material | 2-Methyl-1-[3-(4-morpholinyl)propyl]-5-phenyl-N-[3-(trifluoromethyl)phenyl]-1H-pyrrole-3-carboxamide |
| CAS | 883031-03-6 |
| Quantity | 10 mg |
| Target | TRPV Antagonists |
| Molecular Formula | C26H28F3N3O2 |
| Purity | 0.99 |