Learn More
asecos™ Hazardous Substances Extendable Rack
Product Code. 17866416 Shop All Asecos Products
Click to view available options
Capacity (Metric):
30 L
40 L
Weight (Metric):
49 kg
52 kg
57 kg
64 kg
83 kg
Unit Size:
Each
Product Code. 17866416
Brand: asecos™ 27858
This item requires a price update prior to being ordered.
Request a Price for this item here.
The Hazardous Substances Rack provides storage of aggressive chemicals.
  • Galvanised rack frames
  • Sturdy construction due to reinforcement cross bars.
  • Storage levels in 25 mm increments, height-adjustable.
  • Load capacity per storage shelf: 200 kg (evenly distributed).
  • Flat-pack delivery with assembly instructions
  • Polyethylene sumps with high resistance against aggressive chemicals (such as acids and alkalis).
Specifications
Capacity (English) 7.9 gal.
Capacity (Metric) 30 L
Material Steel
Type Drum Rack
Length (English) 39.84 in.
Length (Metric) 1012.00 mm
Width (English) 39.84 in.
Width (Metric) 1012.00 mm
Height (English) 78.74 in.
Height (Metric) 2000.00 mm
Depth (English) 25.08 in.
Depth (Metric) 637.00 mm
Weight (English) 141.09 lb.
Weight (Metric) 64 kg