Learn More
asecos™ Hazardous Substances Expandable Rack
Product Code. p-200003672 Shop All Asecos Products
Click to view available options
Capacity (Metric):
25 L
30 L
32 L
41 L
45 L
205 L
Unit Size:
Each
Product Code. 17866526
Brand: asecos™ 27842
This item requires a price update prior to being ordered.
Request a Price for this item here.
The Hazardous Substances Rack provides storage of flammable liquids and water pollutants.
  • Galvanised rack frames
  • Sturdy construction due to reinforcement cross bars
  • Storage levels in 25 mm increments, height-adjustable
  • Load capacity per storage shelf: 200 kg (evenly distributed)
  • Flat-pack delivery with assembly instructions
  • Galvanised grids as storage levels and 1 bottom collecting sump made of galvanised steel.
Specifications
Capacity (English) 6.6 gal.
Capacity (Metric) 25 L
Material Steel
Type Drum Rack
Length (English) 51.65 in.
Length (Metric) 1312.00 mm
Width (English) 51.65 in.
Width (Metric) 1312.00 mm
Height (English) 78.74 in.
Height (Metric) 2000.00 mm
Depth (English) 17.20 in.
Depth (Metric) 437.00 mm
Weight (English) 147.71 lb.
Weight (Metric) 67 kg