Learn More
Brady™ Hazardous Chemical Labels for B30 Series
Product Code. 15931011 Shop All Brady Products
Change view
Click to view available options
Display:
L3B Layout
L4A Layout
Unit Size:
Pack of 200
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Unit Size
15931011 L4A Layout Pack of 200
15921011 L3B Layout Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15931011 Supplier Brady™ Supplier No. 121649
  • ONLINE EXCLUSIVE PRICE
    370.00€
    311.59€ / Pack of 200
    Save 58.41€
    15% Off
Estimated Shipment: 18-12-2026
Pre-printed, GHS/CLP chemical labels for BBP3X, S3XXX, and i3300 printers.
  • Permanent acrylic adhesive
  • For smooth and textured surfaces
Specifications
Autoclavable Not Autoclavable
Adhesive Type Permanent Acrylic
Color Red on White
Display L4A Layout
Finish Gloss
Material Vinyl (B-7569)
Print Technology Thermal Transfer
Sterility Non-sterile
For Use With (Application) Facility Identification, Hazard Communication Identification, Electrical Labeling
For Use With (Equipment) BBP™30, BBP™31, BBP™33, BBP™35 Multicolor, BBP™37 Multicolor, BradyPrinter i3300, BradyPrinter S3000, S3100, and i5300 Industrial Label Printers
Labels per Row 1
Quantity 200 Labels
Shape Rectangle
Temperature Range (Metric) -20°C to +75°C
Thickness (Metric) 0.102 mm
Vertical Repeat (Metric) 162 mm
Width (Metric) 155 mm
Height (Metric) 105 mm
Alternate Part Number B30-262-7569-CLP4A