Learn More
Cole-Parmer™ Hayward PVC Needle Valve
Click to view available options
Thread Size:
3/8 in. NPT(F)
Unit Size:
Each
Description
- Accurate flow control lets you adjust flow down to drops per minute
- Seals are heavy-duty FPM O-ring
- Natural PVDF valves are the ideal option for high-purity, high-temperature applications
- Valve body includes flanges for panel mounting
- Connections are standard pipe thread
Specifications
Specifications
| Material | PVC, PTFE, Viton |
| Product Line | Hayward 155.522.554 PVC Needle Valve |
| Inlet | 3/8 in. NPT(F) |
| Outlet Port | 3/8 in. NPT(F) |
| Port Size | 0.375 NPT(F) |
| Thread Size | 3/8 in. NPT(F) |
| Material (Seal) | FPM O-ring |
| Maximum Pressure | 150 psi |
| Max. Operating Temperature (English) | 140°F |
| Max. Operating Temperature (Metric) | 60°C |
| Show More |