Learn More
CHEMetrics Hardness (Total) Titrets Titration Cells Water Testing Kit
Product Code. 11780994
Click to view available options
Range:
100 to 1000 ppm as CaCO3
2 to 20 ppm as CaCO3
20 to 200 ppm as CaCO3
Unit Size:
Pack of 30
Product Code. 11780994
Brand: CHEMetrics K4502
Only null left
Add to basket
Add to basket
Includes
REMOVE
Provides results in two minutes or less
Provide results in two minutes or less
Titrets ampules use reverse titration to quantify concentrations. Each Titret ampule is 13 mm in diameter and is designed for titrimetric analysis. The ampule contains vacuum-sealed liquid titrant and has a flexible valve assembly attached. After snapping the ampule tip, the sample is drawn into the ampule in small does (with the Titrettor™ device included in each kit that precisely controls the sample), (Step 1), until a color change signals that the equivalence point has been reached (Step 2). The titration is stopped at the end point and the ampule is held upright. The liquid level will correspond to a printed scale on the ampule's outer surface (Step 3).
Kits include 30 ampules with valve assemblies, a titrettor, accessory solutions(s) when necessary, a sample cup, and instructions.
Specifications
Analytes Hardness, Total
Includes Cardboard box, 30 ampoules with valve assemblies, titrettor, 25 mL sample cup and instructions
No. of Tests 30
Range 2 to 20 ppm as CaCO3
Test Method Total EDTA
Type Titrets Titration Cells Water Testing Kit
compliance-icons
Product Identifier
  • total hardness titrets ampoules
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P102-Keep out of reach of children.
  • P101-If medical advice is needed, have product container or label at hand.
  • P271-Use only outdoors or in a well-ventilated area.
  • P405-Store locked up.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P103-Read label before use.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P501-Dispose of contents/container to to authorised chemical landfill or if organic to high temperature incineration.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P264-Wash all exposed externalbody areas thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
Supplemental information
  • MIXTURE LIST-Contains : EDTA disodium salt,EDTA magnesium disodium salt, 2-amino-2-ethyl-1,3-propanediol