Learn More
RETSCH Hard Anodized Aluminum Dividing Heads
Brand:  RETSCH 42.793.0001
Additional Details : Weight : 4.60000kg
 View more versions of this product
Product Code. 10044370
  • 4115.00€ / Each
Estimated Shipment: 18-09-2024
Specifications
Dividing Head
For Feed Sizes up to 10mm
8 Quick-release Sample Outlets
Hard Anodized Aluminum