Learn More
Bel-Art™ SP Scienceware™ Handy Stopper/Tubing Size
Product Code. 11392204
Change view
Click to view available options
Material:
Plastic
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material unitSize
11392204 Plastic Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11392204 Supplier Bel-Art™ SP Scienceware™ Supplier No. F133920000
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Easily determines correct size of cork or stopper needed for any flask, bottle
  • Insert thin triangular gauge into vessel and correct stopper size is read at top of neck
  • Two scales provided: one for rubber and vinyl stoppers and the other for corks
  • Back side has circular scale to size tubing outside diameters between 4 and 52mm
Specifications
For Use With (Equipment) Flasks, Bottles
Material Plastic
Product Type Tubing Sizer