Learn More
Brady™ HandiMark™ Vinyl Repositionable Film
Product Code. 10001734
Click to view available options
Color:
White
Yellow
Width (Metric):
13 mm
25 mm
50 mm
Unit Size:
Each
Product Code. 10001734
Brand: Brady™ 042105
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Description HandiMark Repositionable Vinyl Film
Adhesive Type Acrylic, Repositionable
Color Yellow
Finish Gloss
Material Vinyl
Product Type Repositionable Film
For Use With (Application) For temporary, removable and repositionable labels, like 5S and lean inventory labels
For Use With (Equipment) HandiMark
Quantity 1 Roll
Temperature Range (Metric) -40°C to +82°C
Thickness (Metric) 0.031 mm
Length (Metric) 15 m
Width (Metric) 50 mm