missing translation for 'onlineSavingsMsg'
Learn More

TFA Dostmann™ Handheld Anemometer

Product Code. 11759867
Change view
Click to view available options
Type:
Handheld Anemometer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
11759867 Handheld Anemometer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11759867 Supplier TFA Dostmann™ Supplier No. 42.6000.06

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Specifications

Type Handheld Anemometer
Color Blue
For Use With (Application) Measure the current wind strength
Height (Metric) 98 mm
Width (Metric) 39 mm
Temperature (Metric) 30°C to 60°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.