Learn More
asecos™ Hand-Shovel
Product Code. 17876416 Shop All Asecos Products
Change view
Click to view available options
Length (Metric):
187.00 mm
240.00 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
17876416 187.00 mm Each
17866406 240.00 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17876416 Supplier asecos™ Supplier No. 14369
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
The Hand Shovel is designed for spreading and picking up granules.
  • Hand shovel for granules.
  • PP
  • red
Specifications
Material Polypropylene
Type Shovel
Length (English) 7.36 in.
Length (Metric) 187.00 mm
Width (English) 7.36 in.
Width (Metric) 187.00 mm
Height (English) 5.43 in.
Height (Metric) 138.00 mm
Weight (English) 0.42 lb.
Weight (Metric) 0.19 kg