Learn More
Kimberly-Clark™ Scott™ Luxury Foam Frequent Use Hand Cleanser
Product Code. 12621356
Change view
Click to view available options
Volume (Metric):
1000 mL
1200 mL
Unit Size:
Pack of 4
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
12621356 1200 mL Pack of 4
12631376 1000 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12621356 Supplier Kimberly-Clark™ Supplier No. 6345
  • ONLINE EXCLUSIVE PRICE
    184.00€
    41.79€ / Pack of 4
    Save 142.21€
    77% Off
View all promotions
Estimated Shipment: 14-12-2026
Dispenses the ideal amount of soap for each wash preventing overuse. Scott™ Luxury Foam Frequent Use Hand Cleanser delivers up to twice as many washes per liter as conventional liquid soaps.
  • Color-free and fragrance-free foam
  • Luxuriant feel formulated for frequent use
  • Strong, hygienically sealed cassettes that are quick and easy to install
  • Give up to 2,500 shots of soap
  • Cassettes shrink to save space when empty and are recyclable (after the pump is removed)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hand Cleanser
Color Clear
Quantity 4 Electronic Cassettes
Size Up to 2,500 shots
Volume (English) 40.6 oz.
Volume (Metric) 1200 mL
Weight (Metric) 4.8 kg