Learn More
Tork Hand Cleaning Wet Wipe Bucket W14
Shop All Tork Products
Click to view available options
Color:
White
Unit Size:
4 rolls
Description
- Clean hands effectively with these professional-quality wet wipes.
- Avoid potential irritations - these wipes are free of fragrance and kind to your skin.
- Maintain hand hygiene levels when water isn't available with this convenient design.
- Grab wipes easily and efficiently with our bucket dispenser.
Specifications
Specifications
| Product Type | Wet Wipe |
| Length (English) | 17.17 yd. |
| Length (Metric) | 15.7 m |
| Width (English) | 5.31 in. |
| Width (Metric) | 13.5 cm |
| Certifications/Compliance | FSC MIX Credit |
| Color | White |
| Packaging Quantity | 4 x 58 Wipes |
| Ply | 1-ply |
| Certifications/Compliance (Europe) | FSC MIX Credit |