Learn More
Hamilton™ Luer Lock Adapter for Disposable Tips
Product Code. 10005640
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
10005640 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10005640 Supplier Hamilton™ Supplier No. 31331
Estimated Shipment: 02-11-2026
These adapters enable the connection of a standard 1mL or 200μL disposable pipette tip to the end of a Hamilton Syringe.
  • Two luer tip adapters available: 100μL and 1mL
Specifications
Material CTFE
Product Type Luer-tip Adapter
For Use With (Equipment) PB600-1 Repeating Dispensers