Learn More
Hamilton™ 700 Series Microliter™ Shimadzu AOC-14, AOC-17, and AOC-20 Syringes
Click to view available options
Volume (Metric):
10 μL
Unit Size:
Each
Description
Microliter syringe with a stainless steel plunger that is hand-fitted to the glass barrel, designed for liquid sampling applications. Syringe has a removable needle (RN) termination. Available in a 10μL volume
Syringe Termination
- Removable Needle (RN)- The needles are removable and are a Hamilton-specific design. The design allows the needles to seat precisely at the zero graduation mark of the syringe. Users can select the needle gauge, length, and point style to optimize the syringe for custom applications. Additionally, this termination allows for a removable needle without increasing the dead volume of the syringe and is ideal when there is a risk of the needle clogging. Autoclavable when disassembled. Repeated autoclaving will shorten syringe life.
- Point Style 2 - Sharp, beveled, curved, non-coring needle point recommended for septum penetration. Available gauge : 22s
Specifications
Specifications
| Material | Borosilicate Glass (Barrel), Stainless Steel (Plunger) |
| Needle Gauge | 22s G |
| Diameter (English) Inner Needle | 0.005 in. |
| Diameter (Metric) Inner Needle | 0.13 mm |
| Diameter (English) Outer Needle | 0.028 in. |
| Diameter (Metric) Outer Needle | 0.711 mm |
| Length (English) | 1.7 in. |
| Length (English) Needle | 1.69 in. |
| Length (Metric) | 43 mm |
| Product Type | Microliter Syringe |
| Show More |