Learn More
Hamilton™ 1700 Series Gastight™ Syringes
Product Code. 17894212
Click to view available options
Volume (Metric):
50 μL
100 μL
250 μL
500 μL
Needle Gauge:
22 G
22s G
Max. Pressure:
1000 psi
500 psi
700 psi
Model:
1705
1725
1750
Unit Size:
Each
Product Code. 17894212
Brand: Hamilton™ CAL80900
Ideal for dispensing both liquids and gases
  • Calibrated
  • Cemented Needle (N) and Luer Tip Cemented Needle (LTN)
  • 22s/22gauge w/point style-2
  • Gas Sterilizable
  • Precision-machined polymer plunger tip made of PTFE, which provides leak-free seal
  • Gastight fitting enable tip to wipe interior of syringe barrel free of sample
  • Reduces molecule deposition in syringe when samples have dissolved or suspended molecules
  • Volume range: 50μ to 500μL
Needles are 51mm unless user specified
Specifications
Autoclavable Not Autoclavable
Material Borosilicate Glass (Barrel), PTFE (Tip), Stainless Steel (Plunger)
Needle Gauge 22s G
Diameter (English) Inner 0.041 in.
Diameter (English) Inner Needle 0.005 in.
Diameter (Metric) Inner Needle 0.15 mm
Diameter (English) Outer Needle 0.028 in.
Diameter (English) Outer 0.305 in.
Diameter (Metric) Inner 1.03 mm
Diameter (Metric) Outer 7.75 mm
Diameter (Metric) Outer Needle 0.72 mm
For Use With (Application) Ideal for despensing both liquids and gases
Length (English) Needle 2 in.
Product Type Gastight Syringe
Max. Pressure 1000 psi
Max. Temperature (English) 122°F
Max. Temperature (Metric) 50°C
Min. Temperature (English) 50°F
Model 1705
Needle Point Number 2
Min. Temperature (Metric) 10°C
Quantity Ea.
Volume (Metric) 50 μL
Length (Metric) Needle 51 mm
Needle Point Style Sharp, Bevel, Curved, Non-coring
Connection Type Cemented