Learn More
Carl Zeiss™ Halogen Bulbs
Product Code. p-8006874
Click to view available options
Wattage:
37 W
Unit Size:
Each
Product Code. 12201033
Brand: Carl Zeiss™ 0000001879242
This item requires a price update prior to being ordered.
Request a Price for this item here.
Halogen bulb for Carl Zeiss™ educational microscopes
Models with LED illumination are recommended for low temperature, long-term use and have an extremely long service life and low operating costs. Electrical supply 240V, 60Hz, 100W.
  • An educational microscope ideal for teaching
  • Easy to operate, ideal for inexperienced students
  • Robust structure withstands daily wear and tear
  • Quality optics shows the detail and color of any specimen
  • Ergonomic design ideal for long practical sessions
  • Right or left hand operation
  • LED or halogen illumination Suitable for histology, human biology, microbiology, virology and botany.
Specifications
Light Source Halogen
Voltage 12 V
Wattage 37 W
Product Type Halogen Bulb