Learn More
Porcelaines Avignon™ Haldenwanger™ Porcelain Capsule with Flat Bottom
Product Code. 11758433
Click to view available options
Capacity (Metric):
20 mL
22 mL
52 mL
75 mL
90 mL
100 mL
150 mL
200 mL
600 mL
Type:
Capsule with Pouring Spout
Evaporating Capsule
Unit Size:
Pack of 3
Pack of 6
Product Code. 11758433
Brand: Porcelaines Avignon™ 10109
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Use this type of Porcelaines Avignon™ Haldenwanger™ Porcelain Capsule with Flat Bottom in laboratories spanning from educational institutions to scientific development and industrial research..
  • Flat bottom
  • Porcelain crucible
  • White
  • Straight sides
Specifications
Bottom Shape Flat
Capacity (Metric) 150 mL
Material Porcelain
Shape Round
Type Evaporating Capsule
For Use With (Application) Laboratories
Height (Metric) 37 mm
Diameter (Metric) 97 mm