Learn More
Gilac™ HACCP Food Contact Container
Product Code. 17948911
Change view
Click to view available options
Color:
Blue
Green
Red
White
Yellow
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17948911 White Each
17988651 Yellow Each
17978511 Blue Each
17968911 Red Each
17918701 Green Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17948911 Supplier Gilac™ Supplier No. G613221
  • ONLINE EXCLUSIVE PRICE
    50.14€
    35.31€ / Pack of 1
    Save 14.83€
    29% Off
Estimated Shipment: 12-11-2026
HACCP Food Contact Container is ideal for storage, food preparation, and transporting foodstuffs.
  • HACCP round container.
  • Food contact certified.
  • For storage, food preparation and transporting foodstuffs.
  • Highly durable.
  • Reinforced bottom with anti-wear skirt.
  • Reinforced handles to prevent distortion during carrying.
  • Optional, choice of two lids.
  • External Diameter includes handles.
  • Width corresponds to diameter without handles.
Specifications
Product Type HACCP Food Container
Material Polyethylene
Color White
Volume (English) 13.2 gal.
Volume (Metric) 50 L