Learn More
IKA™ HABITAT Ferment, Double-walled 2 L Package
Product Code. 17715089 Shop All IKA Products
Click to view available options
Capacity (Metric):
2 L
Unit Size:
Each
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Product Code. 17715089
missing translation for 'mfr': IKA™ 0010007659
This item requires a price update prior to being ordered.
Request a Price for this item here.
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
The HABITAT ferment package for the bioreactor HABITAT research contains a 2 L double-walled glass vessel for fermentation applications as well as the appropriate stirring drive.
Together with the separately available control tower package (e.g. HABITAT ferment) you receive all necessary components for a successful cultivation. For temperature control via the double jacket we recommend one of our our circulators such as the HRC 2 basic or control.
Scope of delivery
  • Stand 2
  • HA.gv.dw.2 Glass vessel, double-wall
  • HA.mt.s.2 Motor, small
  • HA.hp.s.2 Harvest pipe, straight
  • HA.ino Port for inoculation
  • HA.ba.2 Baffle
  • HA.sp.r.2 Ring sparger, 0.5 mm
  • HA.cn Condenser
  • HA.ip.b.2 6-blade segment impeller
  • HA.s.tm.2 Temperature sensor
  • HA.s.ph.2 pH sensor
  • HA.s.do.2 DO sensor
  • HA.s.fo Foam sensor
  • HA.s.lv.2 Level sensor
  • HA.cab.dw Cable and tube set
  • HA.sf.250 Sample flask (4 pcs.)
TRUSTED_SUSTAINABILITY
Specifications
Type Bioreactor Package
Autoclavable Autoclavable
Capacity (Metric) 2 L
Material Borosilicate Glass
Weight (Metric) 10.55 kg