Learn More
IKA™ Round Bottom Flask Carrier
Product Code. 15265264 Shop All IKA Products
Change view
Click to view available options
Capacity (Metric):
500 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
15265264 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15265264 Supplier IKA™ Supplier No. 0025000639
Compatible with several IKA stirrers.
  • Round/hollow cylindrical bottom
  • Aluminum anodized
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 500 mL
For Use With (Equipment) H135.30 Reaction Block, 500mL, Without Handle
Type H135.30 Reaction Block