Learn More
H-Thr-Leu-OH, TRC
Specifications
Specifications
| Chemical Name or Material | H-Thr-Leu-OH |
| CAS | 50299-12-2 |
| Molecular Formula | C10H20N2O4 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C10H20N2O4/c1-5(2)4-7(10(15)16)12-9(14)8(11)6(3)13/h5-8,13H,4,11H2,1-3H3,(H,12,14)(H,15,16)/t6-,7+,8+/m1/s1 |
| SMILES | CC(C)C[C@H](NC(=O)[C@@H](N)[C@@H](C)O)C(=O)O |
| IUPAC Name | (2S)-2-[[(2S,3R)-2-amino-3-hydroxybutanoyl]amino]-4-methylpentanoic acid |
| Molecular Weight (g/mol) | 232.28 |
| Formula Weight | 232.1423 |