Learn More
Thermo Scientific™ H-Flex 1 Swinging Bucket Rotor and Accessories
Alle ansehen Thermo Scientific Produkte
missing translation for 'orderingAttributeHoverText'
Description:
18 x 15 mL Adapter
33 x 10/12 mL Adapter
400 mL Bottle/Centrilab Adapter
42 x 5/7 mL Adapter
8 x 50 mL Adapter
Biocontainment Lids
Buckets
Cross
Microplate Adapter
Replacement O-Rings
missing translation for 'unitSize'
Each
Pack of 2
Beschreibung
Spin more with less! Whether clinical and diagnostic applications or cell culture and other research, now spin conical tubes, blood tubes and microplates all in one rotor for a small-footprint General Purpose centrifuge. High capacity and flexibility—all in a small footprint.
- Auto-Lock and Click Seal biocontainment lids enable fast, one-handed rotor exchanges and lid locking
- Spins blood tubes, conical tubes and microplates without changing rotor or buckets
- Get results faster without the need to change hardware
- Limit rotor maintenance and contamination potential with slide-coat technology eliminating the need for grease and lubricants
Spezifikation
Spezifikation
| Description | 18 x 15 mL Adapter |
| Material | Molded Polyamide Nylone |
| Type | Swinging Bucket Rotor |
| Capacity | 18 x 15 mL Conical Each Adapter |
| Max. Speed | 4297 |
| For Use With (Equipment) | 75003300 H-Flex Rotor Cross and 75003301 Buckets |
| Biocontainment | Yes |
| Autoclavable | Autoclavable |
| Quantity | Set of 2 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur