Learn More
H-B Instrument™ Enviro-Safe™ General Purpose Liquid-In-Glass Thermometers, Partial Immersion 3 in.
Product Code. p-4880401 Shop All SP Scienceware Products
missing translation for 'orderingAttributeHoverText'
Temperature Range (Metric):
-10°C to +260°C
Scale:
Celsius
missing translation for 'unitSize'
Each
Product Code. 13099132
missing translation for 'mfr': H-B Instrument™ B605030000
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Certified environmentally safe thermometers replace dangerous mercury
  • Earth friendly, EnviroKleen™ certified thermometers
  • Feature green, non-toxic, biodegradable, Enviro-Safe™ liquid against lead-free, white back glass
  • Meet government, education, and industry mandates to purchase “Green” products
Applications:
Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Order Info
Should you require an individualized Thermometer Calibration Report, factory calibrated thermometers or custom calibration services are available.
Certifications
Individually serialized thermometers are traceable to NIST; include a multi-language Statement of Accuracy and Traceability.
Compliance
Manufactured and tested by USA quality team.
Specifications
Certifications/Compliance Individually serialized thermometers are traceable to NIST; include a multi-language Statement of Accuracy and Traceability.
For Use With (Application) Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Immersion Type Partial
Length (English) Overall 14 in.
Length (Metric) Overall 350 mm
Max. Temperature (Metric) 260°C
Min. Temperature (Metric) -10°C
Temperature Range (Metric) -10°C to +260°C
Accuracy ±1.5°C, ±2°C above 100°C, ±2.5°C above 200°C, ±3°C above 230°C
Subdivision 1°C
Immersion Length 76 mm
Scale Celsius