Learn More
H-B Instrument™ Durac™ Compact Sling Psychrometer
Product Code. 17590685
Change view
Click to view available options
Temperature Range (Metric):
-3.88°C to +50°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) Unit Size
17590685 -3.88°C to +50°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17590685 Supplier H-B Instrument™ Supplier No. B615030000
  • ONLINE EXCLUSIVE PRICE
    109.00€
    104.75€ / Pack of 1
    Save 4.25€
    3% Off
View all promotions
 Restricted Item
Estimated Shipment: 20-01-2027
Measures relative humidity with weather bureau-type wet and dry bulb.
  • Organic liquid filled thermometers with white back glass are mounted in a pocket sized self contained plastic case designed with a built in slide rule/convert wet and dry bulb readings/relative humidity, thus eliminating the need for calculating tables
Specifications
Type Compact Sling Psychrometer
Accuracy 1°C
Temperature Range (Metric) -3.88°C to +50°C
Increments 1°
Material Glass
Temperature Range (English) 23°F to 122°F
Length (English) 8.3 in.
Length (Metric) 21 cm
For Use With (Application) To measure relative humidity with weather bureau-type wet and dry bulb
Includes White back glass
Quantity 1 ea.
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • H-B DURAC COMPACT SLING PSYCHROMETER; -5/50C
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P370+P378-In case of fire: Use CO2, powder or waterspray to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contain: acetone