missing translation for 'onlineSavingsMsg'
Learn More

SP Bel-Art™ H-B Durac Plus Pocket Liquid-in-Glass Laboratory Thermometer; Original Liquid Fill

Product Code. 17541295
Change view
Click to view available options
Temperature Range (Metric):
-10°C to 100°C
Accuracy:
±1°C
Includes:
Closed Metal Case
Closed Plastic Case
Window Plastic Case
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) Accuracy Includes unitSize
17541295 -10°C to 100°C ±1°C Closed Metal Case Each
17511295 -10°C to 100°C ±1°C Window Plastic Case Each
17521295 -10°C to 100°C ±1°C Closed Plastic Case Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17541295 Supplier SP Bel-Art™ Supplier No. B607701800

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Featuring a case to protect from breakage, these H-B Durace Plus Pocket Liquid-in-Glass Laboratory Thermometers are a compact 160 mm (6.3 in.), making them ideal for field studies, waste water treatment and any 'on-the-go' applications.

  • Mercury free; contain blue organic liquid against lead-free, white back glass
  • Tested and inspected by our USA quality team
  • Traceable to NIST
  • Include multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability
  • Case options include:
    • Plastic Window - encased in plastic with an open window
    • Closed Plastic - housed in removable plastic case
    • Closed Metal - housed in a removable metal case
  • Should you require an individual Thermometer Calibration Report, factory calibrated thermometers or custom calibration services are available.

Specifications

Immersion Type Total Immersion
Includes Closed Metal Case
Length (Metric) 160 mm
Product Type Thermometer
Length (English) 6.3 in.
Temperature Range (English) 0°F to 220°F
Temperature Range (Metric) -10°C to 100°C
Accuracy ±1°C
compliance-icons compliance-icons
Product Identifier
  • H-B DURAC PLUS POCKET LIQUID-IN-GLASS LAB THERMOME
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P370+P378-In case of fire: Use CO2, powder or waterspray to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contain: acetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.