Learn More
Tocris Bioscience™ H-7 dihydrochloride
GREENER_CHOICE
Product Code. p-7112834
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 18084052
Brand: Tocris Bioscience™ 0542/50
Protein kinase inhibitor
H-7 dihydrochloride is a protein kinase inhibitor (IC50 values for inhibition of PKA, PKG, PKC and myosin light chain kinase are 3.0, 5.8, 6.0 and 97.0 μM, respectively. H-7 shows antiviral activity against influenza A (IC50 = 10 μM) in vitro. H-7 also inhibits interleukin-stimulated secretion of IgM and blocks PMA -stimulated interleukin 1β production.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material '±-1-(5-Isoquinolinesulphonyl)-2-methylpiperazine dihydrochloride
CAS 108930-17-2
Assay Percent Range >99%
Quantity 50 mg
Solubility Soluble to 20mM in DMSO
Molecular Formula C14H17N3O2S.2HCl
Formula Weight Observed MW: 364.29