missing translation for 'onlineSavingsMsg'
Learn More

IKA™ H 66.51 Stainless Steel Spare Sensor

Product Code. 10776913 Shop All IKA Products
Click to view available options
Diameter (Metric):
6 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10776913

Brand: IKA™ 0002735551

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Diameter (Metric) 6 mm
For Use With (Equipment) For ETS-D5
Length (Metric) 260 mm
Model H 66.51
Type Spare Sensor
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.