missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies GW2580
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
- GW2580 is a selective inhibitor of cFMS kinase (IC50 = 0.03 μM) (Conway et al.), 2008), blocking its ability to autophosphorylate colony-stimulating factor (CSF-1 or M-CSF), which promotes the survival, proliferation, and differentiation of macrophages
- Demonstrates the importance of CSF-1 in promoting myeloid lineage bias in mouse hematopoietic stem cells (Mossadegh-Keller et al.)
- Demonstrates the importance of CSF-1 in expansion of mouse and human macrophage colonies and monocytes (He et al.; Clanchy et al.; Conway et al.), 2008; Conway et al.), 2005)
- Inhibits bone degradation in cultures of human osteoclasts, rat calvaria, and rat fetal long bone (Conway et al.), 2005), and in mouse models of arthritis (Conway et al.), 2008)
Specifications
Specifications
| CAS | 870483-87-7 |
| Quantity | 5 mg |
| Molecular Formula | C20H22N4O3 |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction