missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ GW 441756

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Potent, selective inhibitor of the NGF receptor tyrosine kinase A (TrkA) (IC50 = 2 nM). Displays > 100-fold selectivity over a range of other kinases.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 275.31 |
| Purity | >99% |
| Molecular Weight (g/mol) | 275.311 |
| Chemical Name or Material | GW 441756 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction