missing translation for 'onlineSavingsMsg'
Learn More

Gum Acacia, Technical, Powder, (Gum Arabic), Fisher Chemical

Product Code. 10626262 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 10626262

Brand: Fisher Chemical G/1050/53

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

500g, Plastic powder jar

TRUSTED_SUSTAINABILITY

Specifications

Color Beige
pH 7
Physical Form Solid
MDL Number 81264
Quantity 500 g
Packaging Plastic powder jar
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.