Learn More
Checkers Guard Dog™ Low Profile
Product Code. 17752408
Click to view available options
Length (Metric):
27 cm
43 cm
72 cm
77 cm
84 cm
Unit Size:
Each
Product Code. 17752408
Brand: Checkers PGDS0004OB
Only null left
Add to basket
Add to basket
Guard Dog™ Low Profile models are the most compact top-loading cable protectors on the market. These protectors provide a safer crossing for vehicle and pedestrian traffic, while still maintaining a high load-bearing capacity for heavy equipment
  • Guard Dog™ Low Profile models are the most compact top-loading cable protectors on the market
  • These protectors provide a safer crossing for vehicle and pedestrian traffic, while still maintaining a high load-bearing capacity for heavy equipment
Specifications
Length (English) 30.3 in.
Length (Metric) 77 cm
Width (Metric) 91 cm
Color Black, Orange
Material Polyethylene
Type Cable Protector