Learn More
Guanosine 3',5'-Cyclic Monophosphate, TRC
Specifications
Specifications
| Chemical Name or Material | Guanosine 3',5'-Cyclic Monophosphate |
| CAS | 7665-99-8 |
| Molecular Formula | C10 H12 N5 O7 P |
| Quantity | 50 mg |
| Synonym | Guanosine, cyclic 3',5'-(hydrogen phosphate), Guanosine 3',5'-phosphate (cyclic) (7CI), 4H-Furo[3,2-d]-1,3,2-dioxaphosphorin, guanosine deriv., 3',5'-Cyclic GMP, 3',5'-GMP, Cyclic 3',5'-GMP, Cyclic 3',5'-guanylic acid, Cyclic GMP, Cyclic guanosine 3',5'-m |
| InChI Formula | InChI=1S/C10H12N5O7P/c11-10-13-7-4(8(17)14-10)12-2-15(7)9-5(16)6-3(21-9)1-20-23(18,19)22-6/h2-3,5-6,9,16H,1H2,(H,18,19)(H3,11,13,14,17)/t3-,5-,6-,9-/m1/s1 |
| SMILES | NC1=Nc2c(ncn2[C@@H]3O[C@@H]4COP(=O)(O)O[C@H]4[C@H]3O)C(=O)N1 |
| IUPAC Name | 9-[(4aR,6R,7R,7aS)-2,7-dihydroxy-2-oxo-4a,6,7,7a-tetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinin-6-yl]-2-amino-1H-purin-6-one |
| Molecular Weight (g/mol) | 345.205 |
| Formula Weight | 345.047 |