missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanfacine-alpha,alpha-d2 HCl, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Guanfacine-D2 (acetamide-2,2-D2) Hydrochloride |
| CAS | 1398065-88-7 |
| Molecular Formula | C9 D2 H7 Cl2 N3 O . Cl H |
| Quantity | 0.01 g |
| Synonym | N-(Aminoiminomethyl)-2,6-dichlorobenzeneacetamide-α,α-d2 hydrochloride, [(2,6-Dichlorophenyl)acetyl-d2]guanidine hydrochloride, Guanfacine-D2 (acetamide-2,2-D2) hydrochloride, Guanfacine-d2 hydrochloride, N-Amidino-2-(2,6-dichlorophenyl)acetamide-2,2-d2 hydrochloride |
| InChI Formula | InChI=1 S/C9H9Cl2N3O.ClH/c10-6-2-1-3-7(11)5(6)4-8(15)14-9(12)13;/h1-3 H,4H2,(H4,12,13,14,15);1 H/i4D2; |
| SMILES | Cl.[2 H]C([2 H])(C(=O)NC(=N)N)c1c(Cl)cccc1Cl |
| IUPAC Name | N-carbamimidoyl-2,2-dideuterio-2-(2,6-dichlorophenyl)acetamide;hydrochloride |
| Molecular Weight (g/mol) | 284.5665 |
| Formula Weight | 283.0015 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction