missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GSK'872
GREENER_CHOICE

Product Code. 18759786
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18759786

Brand: Tocris Bioscience™ 6492/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective RIP3 kinase inhibitor

GSK'872 is a potent and selective receptor activating protein kinase 3 (RIP3) inhibitor (IC50 = 1.3 nM). Exhibits >1000-fold selectivity for RIP3 over a panel of 300 other kinases including RIP1. Inhibits TNF-induced cell death in 3T3-SA cells and TLR3-induced necrosis in fibroblasts in vitro. Blocks necroptosis of primary human neutrophils from whole blood. Also induces apoptosis at higher concentrations (3 - 10 μM).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-5-Benzothiazolyl-6-[(1-methylethyl)sulfonyl]-4-quinolinamine
CAS 1346546-69-7
Quantity 10 mg
Molecular Formula C19H17N3O2S2
Formula Weight 383.49
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.