Learn More
Tocris Bioscience™ GSK 650394
GREENER_CHOICE
Serum- and glucocorticoid-regulated kinase (SGK) inhibitor
Brand:  Tocris Bioscience™ 3572/10
Encompass
Product Code. 13403224
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
GSK 650394 is a serum- and glucocorticoid-regulated kinase 1 (SGK1) inhibitor (IC50 values are 62 and 103 nM for SGK1 and SGK2 respectively). Displays >30-fold selectivity over Akt and other related kinases. Inhibits androgen-stimulated growth of LNCaP cells, a human prostate carcinoma cell line (IC50 ∽ 1 μM).
Specifications
Specifications
>97%
10 mg