Learn More
Tocris Bioscience™ GSK 650394
Serum- and glucocorticoid-regulated kinase (SGK) inhibitor
Brand: Tocris Bioscience™ 3572/10
Description
GSK 650394 is a serum- and glucocorticoid-regulated kinase 1 (SGK1) inhibitor (IC50 values are 62 and 103 nM for SGK1 and SGK2 respectively). Displays >30-fold selectivity over Akt and other related kinases. Inhibits androgen-stimulated growth of LNCaP cells, a human prostate carcinoma cell line (IC50 ∽ 1 μM).Specifications
| >97% | |
| 10 mg |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only