Learn More
Merck Ground Joint Flasks for Vacuum Filtration
Product Code. p-2429607 Shop All Merck Products
Click to view available options
Volume (Metric):
250 mL
1 L
4 L
5 L
Type:
Conical Bottom Flask
Standard Bottom Flask
Tubing
Unit Size:
Each
Product Code. 15824176
Brand: Merck XX1614705
Only null left
Add to basket
Add to basket
Accessories for filter holders for sample preparation.
  • These accessory products are designed to replace parts contained in Millipore glass vacuum filtration kits or provide for an alternative configuration.
  • This item is an accessory part for the Millipore All-Glass Filter Holders Kits and Millipore MilliSolve Kit.
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Disposable Reusable
For Use With (Equipment) Millipore All-glass Filter Holders Kit, Millipore MilliSolve Kit
Material Borosilicate Glass
Shape Conical Bottom
Type Conical Bottom Flask
Volume (Metric) 1 L