Learn More
Microspec™ Grip Seal Bags
missing translation for 'orderingAttributeHoverText'
Dimensions (L x W):
2 x 9 in.
3 x 7.5 in.
6 x 9 in.
7.5 x 7.5 in.
missing translation for 'unitSize'
Pack of 1000
Description
- Cost effective sampling
- Sample storage of large numbers of samples
Specifications
Specifications
| Content And Storage | Room Temperature |
| Autoclavable | Not Autoclavable |
| Closure Type | Grip Seal |
| Color | Transparent |
| Disposable | Single-use |
| For Use With (Application) | Sampling |
| Material | Polyethylene |
| Sterility | Non-sterile |
| Thickness (Metric) | 50 μm |
| Film Description | Polyethylene |
| Show More |