missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Gremlin 1 Antibody Blocking Peptide
Specifications
Specifications
| Gene ID (Entrez) | 26585 |
| Product Line | Novus |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB100-56883 |
| Gene Alias | Cell proliferation-inducing gene 2 protein, CKTSF1B1gremlin 1, cysteine knot superfamily, homolog, Cysteine knot superfamily 1, BMP antagonist 1 gremlin 1, cysteine knot superfamily, homolog (Xenopus laevis), DAN domain family member 2, DAND2GREMLIN, Down-regulated in Mos-transformed cells protein, DRMMGC126660, gremlin, gremlin 1, gremlin 1-like protein, gremlin-1, IHG-2, Increased in high glucose protein 2, increased in high glucose-2, proliferation-inducing gene 2 |
| Gene Symbol | GREM1 |
| Quantity | 0.1 mg |
| Research Category | Neuroscience |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?