Learn More
Greiner Bio-One Serum Tubes: Clot Activator with Gel Separator
Product Code. 13068682
Click to view available options
Capacity (Metric):
2.5 mL
13 x 100 mm
Closure Color:
Red with White Ring
Red with Yellow Ring
Unit Size:
Pack of 1200
Product Code. 13068682
Brand: Greiner Bio-One 454243
For routine clinical chemistry, immunohematology, serology or immunology testing
  • Coated with micronized silica particles which activate clotting when tubes are gently inverted
  • Tubes with gel contain a barrier gel on the tube bottom that during centrifugation moves upward to the serum-clot interface to form a stable barrier, separating serum from cells
  • Clear as glass with equal or better performance
  • Molded to be virtually unbreakable, providing a better level of safety than glass
  • Tubes have an 18-month shelf life from the date of manufacture
  • Pediatric tubes come with white rings for quick identification
  • Safety Twist Caps can be removed and replaced several times
  • Nonscrew caps can be removed and replaced as often as required
Compliance
Tubes with gel separator are FDA 510(k) cleared for therapeutic drug monitoring (TDM)
TRUSTED_SUSTAINABILITY
Specifications
Additive Microscopic Silica Particles with Separation Gel
Material Plastic
Closure Type Non-ridged Pull Cap
Capacity (Metric) 2.5 mL
Closure Color Red with White Ring