Learn More
Honeywell™ Green Welding Gloves
Product Code. 12610903
Click to view available options
:
Size: 10
Product Code. 12610903
Brand: Honeywell™ 200004110
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Honeywell™ Green Welding Gloves constructed of split leather with a reinforced palm.
  • Split leather gloves, with reinforced palm
  • Gunn cut, sewn-on thumb
  • Yoke around the fingers, thumb, 2nd/3rd finger join, ring finger side and between the cuff and the palm
  • Kevlar™ thread stitching
  • Palm in yellow color, back and cuff in green color
  • Cuff of 140 mm
  • Thickness leather only 1.1/1.3
  • The hide leather is ideal for welders' gloves
  • The US assembly, winged thumb and flexible materials used make this glove anatomical, thus providing excellent comfort for use
  • Split leather treated against the heat for excellent resistance to contact and radiant heat
  • The reinforced palm and thumb in split leather increases the glove's durability
  • The Kevlar™ thread assembly ensures sturdy seams with respect to the heat and wear as well as better thermal insulation
  • The split leather cuff protects a large part of the forearm against mechanical hazards
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 420-03 EEC Basic standards, EN 388-03 Physical Hazards, EN 407-04 Heat hazards, EN 12 477-2000 Protection for Welders Type A
Color Green
Material Leather
Size 10
Length (Metric) 34 cm