Learn More
Honeywell™ Green Welding Gloves
Click to view available options
:
Size: 10
Description
- Split leather gloves, with reinforced palm
- Gunn cut, sewn-on thumb
- Yoke around the fingers, thumb, 2nd/3rd finger join, ring finger side and between the cuff and the palm
- Kevlar™ thread stitching
- Palm in yellow color, back and cuff in green color
- Cuff of 140 mm
- Thickness leather only 1.1/1.3
- The hide leather is ideal for welders' gloves
- The US assembly, winged thumb and flexible materials used make this glove anatomical, thus providing excellent comfort for use
- Split leather treated against the heat for excellent resistance to contact and radiant heat
- The reinforced palm and thumb in split leather increases the glove's durability
- The Kevlar™ thread assembly ensures sturdy seams with respect to the heat and wear as well as better thermal insulation
- The split leather cuff protects a large part of the forearm against mechanical hazards
Specifications
Specifications
| Certifications/Compliance | EN 420-03 EEC Basic standards, EN 388-03 Physical Hazards, EN 407-04 Heat hazards, EN 12 477-2000 Protection for Welders Type A |
| Color | Green |
| Material | Leather |
| Size | 10 |
| Length (Metric) | 34 cm |