Learn More
Deutsch Neumann™ Grey Natural Rubber Stoppers
Click to view available options
Diameter (Metric) Bottom:
3.5 mm
5 mm
8 mm
10.5 mm
12.8 mm
14 mm
17 mm
18 mm
21 mm
23 mm
26 mm
29 mm
31 mm
36 mm
41 mm
47 mm
50.5 mm
56 mm
60 mm
64.5 mm
71 mm
94 mm
Unit Size:
Each
Pack of 25
Description
- Acc. to DIN 12 871
- Durometer 40 Shore A
- Available in a wide range of sizes
Specifications
Specifications
| Height (Metric) | 15mm |
| Quantity | 1/Ea. |
| Color | Gray |
| Diameter (Metric) Bottom | 3.5 mm |
| Diameter (Metric) Top | 6.5 mm |
| Material | Natural Rubber |
| Product Type | Tapered Stopper |