Learn More
Deutsch Neumann™ Grey Natural Rubber Stoppers
Product Code. 10078560
Click to view available options
Diameter (Metric) Bottom:
3.5 mm
5 mm
8 mm
10.5 mm
12.8 mm
14 mm
17 mm
18 mm
21 mm
23 mm
26 mm
29 mm
31 mm
36 mm
41 mm
47 mm
50.5 mm
56 mm
60 mm
64.5 mm
71 mm
94 mm
Unit Size:
Each
Pack of 25
Product Code. 10078560
Brand: Deutsch Neumann™ 1010117
Only null left
Add to basket
Add to basket
For safe and secure closure of laboratory apparatus. Deutsch Neumann™ Grey Natural Rubber Stoppers are suitable for general use.
  • Acc. to DIN 12 871
  • Durometer 40 Shore A
  • Available in a wide range of sizes
Specifications
Height (Metric) 25mm
Quantity 1/Ea.
Color Gray
Diameter (Metric) Bottom 17 mm
Diameter (Metric) Top 22 mm
Material Natural Rubber
Product Type Tapered Stopper