missing translation for 'onlineSavingsMsg'
Learn More

Graveoline, TRC

Product Code. 30391174
Change view
Click to view available options
Quantity:
60 mg
Unit Size:
60mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391174 60 mg 60mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30391174 Supplier Toronto Research Chemicals Supplier No. TRCG78001560MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Graveoline
CAS 485-61-0
Molecular Formula C17H13NO3
Quantity 60 mg
Synonym 2-(1,3-Benzodioxol-5-yl)-1-methyl-4(1H)-quinolinone, Graveolin, Graveoline, NSC 603064, Rutamine
InChI Formula InChI=1S/C17H13NO3/c1-18-13-5-3-2-4-12(13)15(19)9-14(18)11-6-7-16-17(8-11)21-10-20-16/h2-9H,10H2,1H3
SMILES CN1C(=CC(=O)c2ccccc12)c3ccc4OCOc4c3
IUPAC Name 2-(1,3-benzodioxol-5-yl)-1-methylquinolin-4-one
Molecular Weight (g/mol) 279.29
Formula Weight 279.0895
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.